Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REEP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271803.1 | NP_001258732.1 | 7 | 824 | ||
NM_016606.3 | NP_057690.2 | 7 | 818 | |||
NR_073448.1 | 7 | 893 | ||||
NR_073449.1 | 7 | 899 | ||||
GenBank mRNA | AF070551.1 | - | 3 | 262 | ||
AF251041.1 | - | 6 | 598 | |||
AK056193.1 | - | 7 | 780 | |||
AK223611.1 | - | 7 | 765 | |||
AK290547.1 | - | 7 | 714 | |||
AK291101.1 | - | 7 | 777 | |||
AK293480.1 | - | 7 | 792 | |||
AY562240.1 | - | 7 | 610 | |||
BC006218.2 | - | 7 | 732 | 3940709 |