Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REEP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271803.1 | NP_001258732.1 | 7, 8 | 912 | ||
NM_016606.3 | NP_057690.2 | 7, 8 | 906 | |||
NR_073448.1 | 7, 8 | 981 | ||||
NR_073449.1 | 7, 8 | 987 | ||||
GenBank mRNA | AF070551.1 | - | 3, 4 | 350 | ||
AF251041.1 | - | 6, 7 | 686 | |||
AK056193.1 | - | 7, 8 | 868 | |||
AK223611.1 | - | 7, 8 | 853 | |||
AK290547.1 | - | 7, 8 | 802 | |||
AK291101.1 | - | 7, 8 | 865 | |||
AK293480.1 | - | 7, 8 | 880 | |||
AY562240.1 | - | 7, 8 | 698 | |||
BC006218.2 | - | 7, 8 | 820 | 3940709 | ||
BC148256.1 | - | 37 |