Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243266.1 | NP_001230195.1 | 2 | 258 | ||
NM_024021.3 | NP_076926.2 | 3 | 404 | |||
NM_148975.2 | NP_683876.1 | 2 | 258 | |||
XM_017017909.1 | XP_016873398.1 | 2 | 271 | |||
GenBank mRNA | AB013102.1 | - | 2 | 99 | ||
AB022821.1 | - | 2 | 123 | |||
AF068288.1 | - | 2 | 192 | |||
AF237912.1 | - | 2 | 209 | |||
AF350500.1 | - | 1 | 162 | |||
AF354928.1 | - | 1 | 82 | |||
AK075081.1 | - | 3 | 289 | |||
AK292089.1 | - | 2 | 213 | |||
AK314033.1 | - | 2 | 101 | |||
BC020648.1 | - | 2 | 133 | 4720088 | ||
BG538851.1 | - | 2 | 258 | |||
CB046393.1 | - | 2 | 127 |