Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024021.3 | NP_076926.2 | 6 | 771 | ||
NM_148975.2 | NP_683876.1 | 5 | 625 | |||
XM_017017909.1 | XP_016873398.1 | 5 | 638 | |||
GenBank mRNA | AB013102.1 | - | 5 | 466 | ||
AB022821.1 | - | 5 | 490 | |||
AF068288.1 | - | 5 | 559 | |||
AF237912.1 | - | 5 | 576 | |||
AF350500.1 | - | 4 | 529 | |||
AK075081.1 | - | 6 | 656 | |||
AK292089.1 | - | 5 | 580 | |||
AK314033.1 | - | 5 | 468 | |||
BC020648.1 | - | 5 | 500 | 4720088 |