Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015918.3 | NP_057002.2 | 5 | 498 | ||
NM_198202.1 | NP_937845.1 | 4 | 352 | |||
XM_011538441.1 | XP_011536743.1 | 5 | 610 | |||
GenBank mRNA | AA918619.1 | - | 1 | 294 | ||
AF070660.1 | - | 5 | 482 | |||
AF117232.1 | - | 5 | 482 | |||
AJ306296.1 | - | 5 | 485 | |||
AK223206.1 | - | 5 | 470 | |||
AK303144.1 | - | 4 | 333 | |||
BC012505.1 | - | 5 | 485 | 4708484 | ||
BI668578.1 | - | 5 | 505 | |||
BU155588.1 | - | 341 | ||||
CB995722.1 | - | 5 | 620 |