Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015932.5 | NP_057016.1 | 2 | 266 | ||
GenBank mRNA | AF077200.1 | - | 2 | 167 | ||
AF125097.1 | - | 2 | 166 | |||
AF262975.1 | - | 2 | 110 | |||
AF275807.1 | - | 2 | 115 | |||
AK312118.1 | - | 2 | 137 | |||
BC003390.1 | - | 2 | 129 | 3446986 | ||
BC014334.1 | - | 2 | 160 | 4045091 | ||
EF535527.1 | - | 2 | 98 | |||
GU045555.1 | - | 2 | 266 | |||
GU045556.1 | - | 2 | 265 |