Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RWDD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007464.2 | NP_001007465.1 | 8 | 1048 | ||
NM_015952.3 | NP_057036.2 | 7 | 934 | |||
NM_016104.3 | NP_057188.2 | 9 | 1176 | |||
GenBank mRNA | AF092134.1 | - | 7 | 891 | ||
AF132958.1 | - | 7 | 798 | |||
AI129713.1 | - | 1 | 181 | |||
AK290727.1 | - | 7 | 934 | |||
AK301089.1 | - | 8 | 911 | |||
BC015802.1 | - | 7 | 790 | 3909348 | ||
BQ001584.1 | - | 1 | 197 | |||
CA422968.1 | - | 1 | 193 |