Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEMK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317851.1 | NP_001304780.1 | 3 | 599 | ||
NM_016173.4 | NP_057257.1 | 3 | 900 | |||
XM_005265218.3 | XP_005265275.1 | 3 | 900 | |||
XM_011533806.2 | XP_011532108.1 | 3 | 900 | |||
XM_011533807.1 | XP_011532109.1 | 3 | 785 | |||
XM_011533808.1 | XP_011532110.1 | 3 | 571 | |||
XM_011533809.2 | XP_011532111.1 | 3 | 900 | |||
XM_011533810.2 | XP_011532112.1 | 3 | 900 | |||
XM_011533812.2 | XP_011532114.1 | 3 | 900 | |||
XM_011533813.2 | XP_011532115.1 | 3 | 900 | |||
XM_011533814.2 | XP_011532116.1 | 3 | 862 | |||
GenBank mRNA | AF131220.1 | - | 2 | 486 | ||
AF172244.1 | - | 3 | 500 | |||
AK025973.1 | - | 3 | 599 | |||
AK124369.1 | - | 1 | 2218 | |||
AK314023.1 | - | 3 | 520 | |||
BC000781.2 | - | 2 | 468 | 3505225 | ||
BX537562.1 | - | 3 | 536 |