Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191324.1 | NP_001178253.2 | 1 | 202 | ||
NM_016271.4 | NP_057355.2 | 2 | 489 | |||
NM_198128.2 | NP_937761.1 | 1 | 202 | |||
XM_005258286.1 | XP_005258343.1 | 2 | 489 | |||
GenBank mRNA | AF162680.3 | - | 2 | 252 | ||
AK023579.1 | - | 1 | 120 | |||
AK315761.1 | - | 2 | 379 | |||
BC018107.1 | - | 2 | 130 | 3914953 | ||
BI461242.1 | - | 2 | 474 | |||
BI463820.1 | - | 481 | ||||
BM564125.1 | - | 2 | 438 | |||
BT006878.1 | - | 1 | 31 | |||
BU662580.1 | - | 1 | 133 | |||
CR457150.1 | - | 1 | 31 | |||
DA752192.1 | - | 1 | 143 | |||
DB091985.1 | - | 1 | 202 |