Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191324.1 | NP_001178253.2 | 5 | 691 | ||
NM_016271.4 | NP_057355.2 | 6 | 978 | |||
NM_198128.2 | NP_937761.1 | 3 | 409 | |||
XM_005258285.1 | XP_005258342.1 | 5 | 755 | |||
XM_005258286.1 | XP_005258343.1 | 4 | 696 | |||
GenBank mRNA | AF162680.3 | - | 6 | 741 | ||
AK023579.1 | - | 3 | 327 | |||
AK315761.1 | - | 6 | 868 | |||
AL133557.1 | - | 4 | 334 | |||
BC018107.1 | - | 6 | 619 | 3914953 | ||
BT006878.1 | - | 5 | 520 | |||
BU662580.1 | - | 3 | 340 | |||
CR457150.1 | - | 5 | 520 |