Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2J1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016021.2 | NP_057105.2 | 1, 2 | 355 | ||
XM_011535887.1 | XP_011534189.1 | 1, 2 | 342 | |||
XM_011535888.2 | XP_011534190.1 | 1, 2 | 372 | |||
GenBank mRNA | AF151834.1 | - | 1, 2 | 162 | ||
AF161502.1 | - | 1, 2 | 50 | |||
AJ245898.1 | - | 1, 2 | 86 | |||
AK223464.1 | - | 2, 3 | 94 | |||
AK290574.1 | - | 1, 2 | 197 | |||
AK300629.1 | - | 2, 3 | 100 | |||
BC013973.1 | - | 1, 2 | 317 | 4137664 | ||
BF058850.1 | - | 2, 3 | 274 | |||
U93243.1 | - | 1, 2 | 51 |