Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2J1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016021.2 | NP_057105.2 | 2, 3 | 432 | ||
XM_011535887.1 | XP_011534189.1 | 2, 3 | 419 | |||
XM_011535888.2 | XP_011534190.1 | 2, 3 | 449 | |||
GenBank mRNA | AF161502.1 | - | 2, 3 | 127 | ||
AJ245898.1 | - | 2, 3 | 163 | |||
AJ420578.1 | - | 1, 2 | 81 | |||
AK223464.1 | - | 3, 4 | 171 | |||
AK290574.1 | - | 2, 3 | 274 | |||
AK300629.1 | - | 3, 4 | 177 | |||
BC013973.1 | - | 2, 3 | 394 | 4137664 | ||
BF058850.1 | - | 3, 4 | 351 |