Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001260489.1 | NP_001247418.1 | 2 | 588 | ||
NM_001260490.1 | NP_001247419.1 | 2 | 588 | |||
NM_015925.6 | NP_057009.3 | 2 | 588 | |||
NM_205834.3 | NP_991403.1 | 2 | 588 | |||
NM_205835.3 | NP_991404.1 | 2 | 588 | |||
XM_005258980.1 | XP_005259037.1 | 2 | 563 | |||
XM_005258982.1 | XP_005259039.1 | 2 | 563 | |||
XM_011527026.1 | XP_011525328.1 | 2 | 563 | |||
GenBank mRNA | AF130366.1 | - | 2 | 254 | ||
AK075426.1 | - | 2 | 240 | |||
AK126834.1 | - | 2 | 852 | |||
AK130367.1 | - | 2 | 242 | |||
AK296618.1 | - | 2 | 242 | |||
BC000015.2 | - | 2 | 219 | |||
BC004381.1 | - | 2 | 236 | |||
BC047376.1 | - | 2 | 524 | 5272128 | ||
BM837160.1 | - | 2 | 490 | |||
BQ711340.1 | - | 2 | 588 |