Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CES1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003276.2 | 6 | 1110 | |||
GenBank mRNA | AF106005.1 | - | 6 | 1113 | ||
BC131699.1 | - | 5 | 903 |