Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIGD1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271880.1 | NP_001258809.1 | 2 | 199 | ||
NM_016438.3 | NP_057522.1 | 1 | 283 | |||
NR_073504.1 | 1 | 283 | ||||
XM_006721946.2 | XP_006722009.1 | 2 | 177 | |||
XM_006721947.3 | XP_006722010.1 | 2 | 587 | |||
XM_006721948.3 | XP_006722011.1 | 2 | 281 | |||
XM_011524891.2 | XP_011523193.1 | 2 | 681 | |||
XM_017024742.1 | XP_016880231.1 | 2 | 627 | |||
XM_017024743.1 | XP_016880232.1 | 2 | 719 | |||
GenBank mRNA | BC020667.1 | - | 1 | 274 | 4724613 | |
BG573492.1 | - | 1 | 275 | |||
BM503772.1 | - | 1 | 286 | |||
BM876435.1 | - | 1 | 259 | |||
BQ017345.1 | - | 3 | 359 |