Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALML5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017422.4 | NP_059118.2 | 1 | 282 | ||
GenBank mRNA | AF172852.1 | - | 1 | 263 | ||
AK222744.1 | - | 1 | 282 | |||
BC039172.1 | - | 1 | 267 | 4753280 |