Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHIC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039840.2 | NP_001034929.2 | 6 | 2681 | ||
NM_001300884.1 | NP_001287813.1 | 5 | 2621 | |||
XM_017029583.1 | XP_016885072.1 | 7 | 2362 | |||
GenBank mRNA | AL832095.1 | - | 6 | 2321 | ||
BC136691.1 | - | 6 | 2586 | 9020681 | ||
BC136692.1 | - | 6 | 2586 | 9020682 | ||
CR936642.1 | - | 5 | 2572 |