Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM6SF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001524.2 | NP_001001524.2 | 5 | 516 | ||
GenBank mRNA | AF255923.1 | - | 5 | 465 | ||
AK026307.1 | - | 3 | 925 | |||
AK097070.1 | - | 3 | 987 | |||
BC120986.1 | - | 5 | 490 | |||
BC120987.1 | - | 5 | 490 |