Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFYVE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281734.1 | NP_001268663.1 | 5 | 1998 | ||
NM_001281735.1 | NP_001268664.1 | 2 | 134 | |||
NM_021260.3 | NP_067083.1 | 5 | 1998 | |||
NM_178441.2 | NP_848535.1 | 2 | 139 | |||
XM_017021373.1 | XP_016876862.1 | 3 | 663 | |||
GenBank mRNA | AB046809.1 | - | 5 | 1823 | ||
AF251025.2 | - | 4 | 1703 | |||
AF311602.1 | - | 2 | 139 | |||
AF318319.1 | - | 2 | 166 | |||
AJ310569.1 | - | 5 | 1849 | |||
AK027399.1 | - | 5 | 1892 | |||
AK098542.1 | - | 2 | 122 | |||
BC014902.1 | - | 5 | 1917 | |||
BC053520.1 | - | 5 | 1926 | 5493027 |