Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFYVE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281735.1 | NP_001268664.1 | 5 | 401 | ||
NM_021260.3 | NP_067083.1 | 8 | 2265 | |||
NM_178441.2 | NP_848535.1 | 5 | 406 | |||
XM_017021373.1 | XP_016876862.1 | 6 | 930 | |||
GenBank mRNA | AB046809.1 | - | 8 | 2090 | ||
AF251025.2 | - | 7 | 1970 | |||
AF311602.1 | - | 5 | 406 | |||
AF318319.1 | - | 5 | 434 | |||
AJ310569.1 | - | 8 | 2116 | |||
AK027399.1 | - | 8 | 2159 | |||
AK098542.1 | - | 5 | 389 | |||
BC053520.1 | - | 8 | 2193 | 5493027 |