Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFYVE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281734.1 | NP_001268663.1 | 7, 8 | 2249 | ||
NM_001281735.1 | NP_001268664.1 | 5 | 427 | |||
NM_021260.3 | NP_067083.1 | 8 | 2291 | |||
NM_178441.2 | NP_848535.1 | 5 | 432 | |||
XM_017021373.1 | XP_016876862.1 | 6 | 956 | |||
GenBank mRNA | AB046809.1 | - | 8 | 2116 | ||
AF251025.2 | - | 7 | 1996 | |||
AF311602.1 | - | 5 | 432 | |||
AF318319.1 | - | 5 | 460 | |||
AJ310569.1 | - | 8 | 2142 | |||
AK027399.1 | - | 8 | 2185 | |||
AK098542.1 | - | 5 | 415 | |||
BC014902.1 | - | 7, 8 | 2168 | |||
BC053520.1 | - | 8 | 2219 | 5493027 |