Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FXYD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184963.1 | NP_001171892.1 | 5 | 278 | ||
NM_173160.2 | NP_775183.1 | 5 | 414 | |||
GenBank mRNA | AA759244.1 | - | 5 | 358 | ||
AI829935.1 | - | 5 | 365 | |||
AY358584.1 | - | 5 | 278 | |||
BC054876.1 | - | 5 | 420 | 4610529 | ||
BG432244.1 | - | 5 | 417 |