Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YBEY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006114.2 | NP_001006114.1 | 3 | 388 | ||
NM_001314023.1 | NP_001300952.1 | 3 | 253 | |||
NM_001314025.1 | NP_001300954.1 | 3 | 388 | |||
NM_001314026.1 | NP_001300955.1 | 3 | 710 | |||
NM_058181.2 | NP_478061.1 | 3 | 710 | |||
XM_011529629.2 | XP_011527931.1 | 3 | 358 | |||
XM_011529630.2 | XP_011527932.1 | 3 | 389 | |||
XM_011529631.2 | XP_011527933.1 | 3 | 358 | |||
XM_011529633.2 | XP_011527935.1 | 3 | 358 | |||
XM_011529635.2 | XP_011527937.1 | 3 | 389 | |||
XM_017028396.1 | XP_016883885.1 | 405 | ||||
GenBank mRNA | AI262212.1 | - | 3 | 339 | ||
AK128875.1 | - | 3 | 322 | |||
AK294975.1 | - | 3 | 369 | |||
AY040873.1 | - | 3 | 656 | |||
AY040875.1 | - | 3 | 199 | |||
BC022828.1 | - | 3 | 329 | 3928883 | ||
BC119770.1 | - | 3 | 334 | 40108760 | ||
BC122865.1 | - | 3 | 334 | 40108759 | ||
BC131553.1 | - | 3 | 334 | 40089560 | ||
BG473705.1 | - | 2 | 294 | |||
BM796288.1 | - | 3 | 381 |