Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018989.1 | NP_061862.1 | 11 | 1806 | ||
XM_005268466.1 | XP_005268523.1 | 11 | 1859 | |||
XM_005268467.1 | XP_005268524.1 | 10 | 1694 | |||
XM_005268468.1 | XP_005268525.1 | 10 | 1691 | |||
XM_006714787.1 | XP_006714850.1 | 12 | 1823 | |||
XM_011537661.1 | XP_011535963.1 | 11 | 1859 | |||
XM_011537662.1 | XP_011535964.1 | 11 | 1856 | |||
XM_017009587.1 | XP_016865076.1 | 12 | 1820 | |||
GenBank mRNA | AB037732.1 | - | 7 | 1129 | ||
AK128738.1 | - | 10 | 1691 | |||
BX647384.1 | - | 11 | 1806 |