Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005354.2 | NP_001005354.1 | 3 | 242 | ||
NM_018457.3 | NP_060927.1 | 3 | 392 | |||
GenBank mRNA | AF217517.1 | - | 1 | 305 | ||
AK074398.1 | - | 1 | 288 | |||
BC001464.2 | - | 3 | 378 | |||
BC014257.2 | - | 1 | 292 | 4765821 | ||
BC016064.1 | - | 1 | 289 | 4852862 | ||
BC021019.1 | - | 1 | 260 | |||
BC071979.1 | - | 1 | 235 | |||
BX640962.1 | - | 2 | 130 | |||
CN392914.1 | - | 3 | 406 | |||
CR533459.1 | - | 2 | 228 |