Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXW5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018998.3 | NP_061871.1 | 6 | 1087 | ||
XM_005266089.2 | XP_005266146.2 | 6 | 1137 | |||
XM_011518789.1 | XP_011517091.1 | 6 | 1137 | |||
XM_017014812.1 | XP_016870301.1 | 6 | 1107 | |||
GenBank mRNA | AB209726.1 | - | 3 | 499 | ||
AF217998.1 | - | 2 | 249 | |||
AF258569.1 | - | 2 | 248 | |||
AK026081.1 | - | 6 | 951 | |||
AK315738.1 | - | 6 | 962 | |||
AL137631.1 | - | 2 | 350 | |||
BC004541.2 | - | 3 | 496 | |||
BC009429.1 | - | 1 | 240 | |||
BC014130.1 | - | 6 | 983 | 4620371 | ||
BC014297.2 | - | 6 | 954 | 3958395 | ||
BC031100.1 | - | 5 | 1102 | |||
BC035355.1 | - | 5 | 1101 |