Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXW5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018998.3 | NP_061871.1 | 8 | 1593 | ||
XM_005266089.2 | XP_005266146.2 | 8 | 1643 | |||
XM_011518789.1 | XP_011517091.1 | 8 | 1589 | |||
XM_017014812.1 | XP_016870301.1 | 8 | 1559 | |||
GenBank mRNA | AB209726.1 | - | 5 | 1005 | ||
AF217998.1 | - | 4 | 753 | |||
AF258569.1 | - | 4 | 751 | |||
AK026081.1 | - | 8 | 1457 | |||
AK315738.1 | - | 8 | 1468 | |||
AL137631.1 | - | 4 | 1147 | |||
BC000850.1 | - | 2 | 199 | |||
BC004541.2 | - | 5 | 1002 | |||
BC009429.1 | - | 3 | 746 | |||
BC014130.1 | - | 8 | 1489 | 4620371 | ||
BC014297.2 | - | 8 | 1460 | 3958395 | ||
BC031100.1 | - | 7 | 1554 | |||
BC035355.1 | - | 7 | 1607 |