Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKIB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019004.1 | NP_061877.1 | 3 | 584 | ||
XM_005250458.3 | XP_005250515.1 | 3 | 602 | |||
XM_011516334.1 | XP_011514636.1 | 3 | 602 | |||
XM_011516335.2 | XP_011514637.1 | 3 | 166 | |||
XM_017012359.1 | XP_016867848.1 | 3 | 602 | |||
GenBank mRNA | AB037807.1 | - | 3 | 584 | ||
BC063861.1 | - | 3 | 560 |