Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFAND6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242911.1 | NP_001229840.1 | 5, 6 | 727 | ||
NM_001242912.1 | NP_001229841.1 | 6, 7 | 806 | |||
NM_001242913.1 | NP_001229842.1 | 7, 8 | 761 | |||
NM_001242914.1 | NP_001229843.1 | 6, 7 | 736 | |||
NM_001242915.1 | NP_001229844.1 | 5, 6 | 572 | |||
NM_001242916.1 | NP_001229845.1 | 6, 7 | 735 | |||
NM_001242917.1 | NP_001229846.1 | 6, 7 | 735 | |||
NM_001242919.1 | NP_001229848.1 | 5, 6 | 567 | |||
NM_019006.3 | NP_061879.2 | 6, 7 | 890 | |||
XM_011521684.2 | XP_011519986.1 | 7, 8 | 773 | |||
GenBank mRNA | AF061739.1 | - | 6, 7 | 675 | ||
AF261138.1 | - | 5, 6 | 561 | |||
AJ251095.1 | - | 5, 6 | 524 | |||
AK055881.1 | - | 1, 2 | 1684 | |||
AL136598.1 | - | 5, 6 | 582 | |||
BC005283.1 | - | 6, 7 | 779 | 4042180 | ||
CN428457.1 | - | 761 | ||||
CR533476.1 | - | 4, 5 | 468 | |||
DR762914.1 | - | 6, 7 | 720 |