Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM64A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195228.1 | NP_001182157.1 | 2 | 215 | ||
NM_019013.2 | NP_061886.2 | 2 | 215 | |||
XM_011523944.2 | XP_011522246.1 | 2 | 325 | |||
GenBank mRNA | AK001018.1 | - | 1 | 155 | ||
AK001353.1 | - | 2 | 175 | |||
AK225613.1 | - | 2 | 177 | |||
AK225702.1 | - | 2 | 175 | |||
BC005004.1 | - | 2 | 182 | 3533875 | ||
BC013966.2 | - | 2 | 161 | 2988318 | ||
KU178653.1 | - | 1 | 106 | |||
KU178654.1 | - | 1 | 106 |