Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMCO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256164.1 | NP_001243093.1 | 2 | 187 | ||
NM_001256165.1 | NP_001243094.1 | 2 | 489 | |||
NM_019026.4 | NP_061899.2 | 2 | 289 | |||
NR_045818.1 | 2 | 212 | ||||
GenBank mRNA | AB020980.1 | - | 2 | 256 | ||
AF070626.1 | - | 2 | 245 | |||
AF274935.1 | - | 2 | 125 | |||
AF277194.1 | - | 2 | 163 | |||
AK298614.1 | - | 2 | 456 | |||
AK316080.1 | - | 2 | 422 | |||
AK316610.1 | - | 2 | 235 | |||
AY359027.1 | - | 2 | 224 | |||
AY763589.1 | - | 2 | 218 | |||
BC000104.2 | - | 2 | 209 | 3504808 | ||
BG772940.1 | - | 2 | 294 | |||
BP257387.1 | - | 2 | 185 | |||
CN430121.1 | - | 2 | 267 |