Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009941.2 | NP_001009941.1 | 7 | 1534 | ||
NM_001009943.2 | NP_001009943.1 | 6 | 1455 | |||
NM_019046.2 | NP_061919.1 | 7 | 1534 | |||
GenBank mRNA | AA431197.1 | - | 1 | 146 | ||
AI738771.1 | - | 1 | 131 | |||
AL137614.1 | - | 4 | 389 | |||
AW470888.1 | - | 1 | 133 | |||
BC062346.1 | - | 7 | 1020 | 3845142 |