Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009941.2 | NP_001009941.1 | 3, 4 | 1117 | ||
NM_001009943.2 | NP_001009943.1 | 3, 4 | 1117 | |||
NM_019046.2 | NP_061919.1 | 3, 4 | 1117 | |||
GenBank mRNA | AI738771.1 | - | 3, 4 | 469 | ||
AW470888.1 | - | 4, 5 | 445 | |||
AW630443.1 | - | 2, 3 | 196 | |||
BC062346.1 | - | 3, 4 | 603 | 3845142 |