Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019050.2 | NP_061923.2 | 15 | 3152 | ||
XM_005263073.3 | XP_005263130.1 | 14 | 3026 | |||
XM_005263075.2 | XP_005263132.1 | 14 | 3041 | |||
XM_005263076.2 | XP_005263133.1 | 14 | 3038 | |||
XM_005263077.3 | XP_005263134.1 | 15 | 3191 | |||
XM_006714240.1 | XP_006714303.1 | 13 | 2227 | |||
XM_011532037.2 | XP_011530339.1 | 15 | 3204 | |||
XM_011532038.2 | XP_011530340.1 | 14 | 2481 | |||
XM_017008312.1 | XP_016863801.1 | 16 | 3318 | |||
XM_017008313.1 | XP_016863802.1 | 15 | 3003 | |||
XM_017008314.1 | XP_016863803.1 | 15 | 3002 | |||
XM_017008315.1 | XP_016863804.1 | 15 | 3072 | |||
XM_017008316.1 | XP_016863805.1 | 13 | 2890 | |||
XM_017008317.1 | XP_016863806.1 | 14 | 3015 | |||
XM_017008318.1 | XP_016863807.1 | 15 | 3137 | |||
GenBank mRNA | AB037771.1 | - | 8 | 1601 | ||
AJ583824.1 | - | 12 | 2083 | |||
AK025301.1 | - | 4 | 1053 | |||
BX648339.1 | - | 13 | 2240 | |||
CR749490.1 | - | 16 | 3268 |