Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM35A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019054.2 | NP_061927.2 | 7, 8 | 2317 | ||
XM_005269920.3 | XP_005269977.1 | 8, 9 | 1467 | |||
XM_006717903.3 | XP_006717966.1 | 9, 10 | 2658 | |||
XM_006717905.3 | XP_006717968.1 | 8, 9 | 2581 | |||
XM_011539876.2 | XP_011538178.1 | 7, 8 | 2508 | |||
XM_011539877.2 | XP_011538179.1 | 10, 11 | 2723 | |||
XM_011539878.2 | XP_011538180.1 | 8, 9 | 2581 | |||
XM_011539879.2 | XP_011538181.1 | 7, 8 | 2567 | |||
XM_011539881.2 | XP_011538183.1 | 6, 7 | 2352 | |||
XM_011539882.2 | XP_011538184.1 | 6, 7 | 2301 | |||
XM_011539883.2 | XP_011538185.1 | 7, 8 | 1512 | |||
XM_017016349.1 | XP_016871838.1 | 12, 13 | 2967 | |||
XM_017016350.1 | XP_016871839.1 | 9, 10 | 2926 | |||
XM_017016351.1 | XP_016871840.1 | 8, 9 | 2514 | |||
XM_017016352.1 | XP_016871841.1 | 8, 9 | 2463 | |||
XM_017016353.1 | XP_016871842.1 | 7, 8 | 1260 | |||
GenBank mRNA | AF131775.1 | - | 5, 6 | 826 | ||
AK022978.1 | - | 7, 8 | 2317 | |||
AK025606.1 | - | 5, 6 | 1166 | |||
BC051863.2 | - | 6, 7 | 2263 | 6146593 |