Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019056.6 | NP_061929.2 | 2, 3 | 892 | ||
GenBank mRNA | AF251063.1 | - | 2, 3 | 725 | ||
BC107805.1 | - | 2, 3 | 375 | 6496406 | ||
BU682304.1 | - | 1, 2 | 246 |