Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ING3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019071.2 | NP_061944.2 | 2, 3 | 237 | ||
NM_198267.1 | NP_938008.1 | 2, 3 | 237 | |||
XM_017012369.1 | XP_016867858.1 | 2, 3 | 245 | |||
GenBank mRNA | AF074968.1 | - | 2, 3 | 214 | ||
AF161419.1 | - | 2, 3 | 227 | |||
AK000096.1 | - | 2, 3 | 238 | |||
AK291905.1 | - | 2, 3 | 215 | |||
AK293682.1 | - | 1, 2 | 123 | |||
AL603623.1 | - | 2, 3 | 214 | |||
AY007790.1 | - | 2, 3 | 218 | |||
BC009776.1 | - | 2, 3 | 192 | |||
BC009777.1 | - | 2, 3 | 225 | |||
BC010851.1 | - | 2, 3 | 184 | |||
BC062634.1 | - | 2, 3 | 200 | |||
BC073865.1 | - | 2, 3 | 205 | |||
BC093091.1 | - | 2, 3 | 221 | |||
BC093689.1 | - | 2, 3 | 197 | |||
BC101609.1 | - | 2, 3 | 197 | |||
BC143316.1 | - | 2, 3 | 197 |