Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHB18P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_001281.2 | 1 | 1115 | |||
GenBank mRNA | AF152528.1 | - | LIT2076 PCDH-psi2 |
1 | 897 | |
AF217743.1 | - | 1 | 734 | |||
AK095496.1 | - | 1 | 1084 |