Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTUD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102653.1 | NP_001096123.1 | 2, 3 | 193 | ||
NM_017493.6 | NP_059963.1 | 1, 2 | 91 | |||
XM_005263079.4 | XP_005263136.1 | 2, 3 | 296 | |||
GenBank mRNA | AK302581.1 | - | 2, 3 | 155 | ||
BC118572.1 | - | 1, 2 | 87 | 40036249 | ||
BC118653.1 | - | 1, 2 | 87 | 40036248 | ||
DB446926.2 | - | 2, 3 | 195 | |||
X68242.1 | - | 2, 3 | 182 |