Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMED9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017510.4 | NP_059980.2 | 1 | 140 | ||
GenBank mRNA | AF441399.2 | - | 1 | 92 | ||
AK298435.1 | - | 1 | 80 | |||
AK313292.1 | - | 1 | 149 | |||
BC001123.2 | - | 1 | 140 | 2967520 | ||
X90872.1 | - | 1 | 165 |