Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMED9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017510.4 | NP_059980.2 | 1 | 172 | ||
GenBank mRNA | AF441399.2 | - | 1 | 124 | ||
AK298435.1 | - | 1 | 112 | |||
AK313292.1 | - | 1 | 181 | |||
BC001123.2 | - | 1 | 172 | 2967520 | ||
X90872.1 | - | 1 | 197 |