Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DEF8 SIRNA Assays ›
Gene Symbol:
DEF8
Gene Name:
differentially expressed in FDCP 8 homolog (mouse)
Gene Aliases:
-
Chromosome Location:
Chr.16: 89948697 - 89968060 on Build GRCh38
UniGene ID:
Hs.62771
Species:
Human
siRNA ID
HSS147794
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DEF8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242816.1
NP_001229745.1
6
617
NM_001242817.1
NP_001229746.1
5
513
NM_001242818.1
NP_001229747.1
6
647
NM_001242819.1
NP_001229748.1
5
560
NM_001242820.1
NP_001229749.1
7
887
NM_001242821.1
NP_001229750.1
5
550
NM_001242822.1
NP_001229751.1
6
644
NM_017702.3
NP_060172.1
6
647
NM_207514.2
NP_997397.1
6
772
XM_005256318.2
XP_005256375.2
6
623
XM_011523156.2
XP_011521458.1
6
623
XM_011523157.2
XP_011521459.1
6
623
XM_011523158.1
XP_011521460.1
6
623
XM_011523159.2
XP_011521461.1
6
571
XM_011523160.2
XP_011521462.1
6
724
XM_011523161.2
XP_011521463.1
6
705
XM_011523162.2
XP_011521464.1
5
584
XM_017023358.1
XP_016878847.1
6
623
XM_017023359.1
XP_016878848.1
6
623
XM_017023360.1
XP_016878849.1
6
623
XM_017023361.1
XP_016878850.1
6
571
XM_017023362.1
XP_016878851.1
6
571
XM_017023363.1
XP_016878852.1
6
705
XM_017023364.1
XP_016878853.1
6
571
XM_017023365.1
XP_016878854.1
6
724
XM_017023366.1
XP_016878855.1
5
1688
XM_017023367.1
XP_016878856.1
5
584
XM_017023368.1
XP_016878857.1
5
1688
XM_017023369.1
XP_016878858.1
5
584
GenBank mRNA
AK000193.1
-
6
583
AK095051.1
-
6
728
AK096485.1
-
4
208
AK131370.1
-
6
553
AK225539.1
-
6
588
AK295880.1
-
7
849
AK296410.1
-
5
449
AK303510.1
-
5
494
BC015482.2
-
5
570
BC105592.1
-
5
490
BC130482.1
-
5
533
BC144038.1
-
5
533
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
intracellular signal transduction
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0