Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM86C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099653.1 | NP_001093123.1 | 4 | 357 | ||
NM_018172.2 | NP_060642.2 | 4 | 378 | |||
NM_152563.2 | NP_689776.1 | 3 | 276 | |||
GenBank mRNA | AK001523.1 | - | 4 | 352 | ||
AK130709.1 | - | 3 | 273 | |||
AK293221.1 | - | 3 | 250 | |||
AK311428.1 | - | 4 | 334 | |||
AK311518.1 | - | 4 | 357 | |||
AK311675.1 | - | 5 | 463 | |||
AK315454.1 | - | 3 | 250 | |||
BC032519.1 | - | 3 | 276 | 5494088 | ||
BQ921333.1 | - | 4 | 327 | |||
DC312827.1 | - | 5 | 462 |
Gene Symbol: ALG1L9P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073388.1 | 4 | 711 | |||
GenBank mRNA | AL534280.3 | - | 5 | 786 |