Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHDC8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271863.1 | NP_001258792.1 | 5, 6 | 1233 | ||
NM_001271864.1 | NP_001258793.1 | 3, 4 | 693 | |||
NM_001271865.1 | NP_001258794.1 | 4, 5 | 908 | |||
NM_018203.2 | NP_060673.1 | 2, 3 | 1021 | |||
GenBank mRNA | AK001610.1 | - | 2, 3 | 932 | ||
AK096590.1 | - | 5, 6 | 1182 | |||
BC036076.1 | - | 2, 3 | 602 | 5302460 |