Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM206 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198862.1 | NP_001185791.1 | 7 | 1050 | ||
NM_018252.2 | NP_060722.2 | 6 | 891 | |||
XM_005273177.3 | XP_005273234.1 | 6 | 1351 | |||
XM_011509717.2 | XP_011508019.1 | 6 | 1150 | |||
XM_017001634.1 | XP_016857123.1 | 5 | 965 | |||
XM_017001635.1 | XP_016857124.1 | 5 | 1264 | |||
XM_017001636.1 | XP_016857125.1 | 5 | 734 | |||
GenBank mRNA | AK001736.1 | - | 6 | 861 | ||
AK024066.1 | - | 6 | 869 | |||
AK295224.1 | - | 6 | 881 | |||
AK297200.1 | - | 7 | 1050 | |||
BC006320.2 | - | 6 | 850 | 4137890 | ||
CR457268.1 | - | 6 | 723 |