Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIDD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145886.3 | NP_665893.2 | 5, 6 | 1115 | ||
NM_145887.3 | NP_665894.2 | 5, 6 | 1115 | |||
XM_005253005.4 | XP_005253062.1 | 6, 7 | 2053 | |||
XM_005253006.4 | XP_005253063.1 | 6, 7 | 1616 | |||
XM_005253007.3 | XP_005253064.1 | 5, 6 | 1133 | |||
XM_005253008.4 | XP_005253065.1 | 6, 7 | 2113 | |||
XM_011520209.2 | XP_011518511.1 | 5, 6 | 1351 | |||
XM_011520210.2 | XP_011518512.1 | 6, 7 | 2053 | |||
XM_011520211.2 | XP_011518513.1 | 6, 7 | 2053 | |||
XM_011520212.1 | XP_011518514.1 | 5, 6 | 1193 | |||
XM_011520213.2 | XP_011518515.1 | 3, 4 | 335 | |||
XM_017017993.1 | XP_016873482.1 | 2, 3 | 547 | |||
GenBank mRNA | AB208832.1 | - | 3, 4 | 1919 | ||
AB208949.1 | - | 3, 4 | 1883 | |||
AB209529.1 | - | 3, 4 | 2025 | |||
AF229178.1 | - | 4, 5 | 1317 | |||
AF274972.1 | - | 5, 6 | 1111 | |||
AF465246.1 | - | 2, 3 | 495 | |||
AK074893.1 | - | 2, 3 | 118 | |||
AL833849.1 | - | 2, 3 | 351 | |||
BC014904.1 | - | 4, 5 | 1053 | 3908102 | ||
BQ924290.1 | - | 2, 3 | 388 | |||
BX349115.2 | - | 3, 4 | 468 |