Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIDD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145886.3 | NP_665893.2 | 4 | 909 | ||
NM_145887.3 | NP_665894.2 | 4 | 909 | |||
XM_005253005.4 | XP_005253062.1 | 5 | 1847 | |||
XM_005253006.4 | XP_005253063.1 | 5 | 1410 | |||
XM_005253007.3 | XP_005253064.1 | 4 | 927 | |||
XM_005253008.4 | XP_005253065.1 | 5 | 1907 | |||
XM_011520209.2 | XP_011518511.1 | 4 | 1145 | |||
XM_011520210.2 | XP_011518512.1 | 5 | 1847 | |||
XM_011520211.2 | XP_011518513.1 | 5 | 1847 | |||
XM_011520212.1 | XP_011518514.1 | 4 | 987 | |||
XM_011520213.2 | XP_011518515.1 | 2 | 129 | |||
XM_017017993.1 | XP_016873482.1 | 1 | 341 | |||
GenBank mRNA | AB208832.1 | - | 2 | 1713 | ||
AB208949.1 | - | 2 | 1677 | |||
AB209529.1 | - | 2 | 1819 | |||
AF229178.1 | - | 3 | 1111 | |||
AF274972.1 | - | 4 | 905 | |||
AF465246.1 | - | 1 | 289 | |||
AL833849.1 | - | 1 | 145 | |||
BC014904.1 | - | 3 | 847 | 3908102 | ||
BX349115.2 | - | 2 | 262 |