Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136037.2 | NP_001129509.2 | 11 | 1162 | ||
NM_001161403.1 | NP_001154875.1 | 10 | 1096 | |||
NM_001161404.1 | NP_001154876.1 | 10 | 1386 | |||
NM_001256542.1 | NP_001243471.1 | 7 | 788 | |||
NM_017980.4 | NP_060450.2 | 10 | 1207 | |||
XM_005263710.2 | XP_005263767.1 | 9 | 879 | |||
XM_006712627.3 | XP_006712690.1 | 7 | 688 | |||
XM_011511453.1 | XP_011509755.1 | 9 | 943 | |||
XM_017004469.1 | XP_016859958.1 | 5 | 653 | |||
GenBank mRNA | AF484961.1 | - | 9 | 930 | ||
AF520987.1 | - | 10 | 1551 | |||
AF527764.1 | - | 10 | 1161 | |||
AF527765.1 | - | 10 | 1023 | |||
AF527766.1 | - | 10 | 1063 | |||
AF527767.1 | - | 9 | 1319 | |||
AF527768.1 | - | 8 | 678 | |||
AF527769.1 | - | 6 | 752 | |||
AF527770.1 | - | 7 | 715 | |||
AK000906.1 | - | 8 | 578 | |||
AK022470.1 | - | 10 | 975 | |||
AK091652.1 | - | 8 | 1457 | |||
AK093692.1 | - | 4 | 1150 | |||
AK094954.1 | - | 9 | 1643 | |||
AK095790.1 | - | 13 | 1611 | |||
AK123014.1 | - | 5 | 3855 | |||
AK297645.1 | - | 12 | 1354 | |||
AL512716.1 | - | 1 | 35 | |||
BC001370.2 | - | 2 | 186 | |||
BC008977.1 | - | 2 | 101 | |||
BC065816.1 | - | 10 | 1014 | 6181259 | ||
BX458594.2 | - | 7 | 744 |