Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ENAH SIRNA Assays ›
Gene Symbol:
ENAH
Gene Name:
enabled homolog (Drosophila)
Gene Aliases:
ENA, MENA, NDPP1
Chromosome Location:
Chr.1: 225486832 - 225660045 on Build GRCh38
UniGene ID:
Hs.497893
Species:
Human
siRNA ID
HSS148098
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ENAH
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001008493.1
NP_001008493.1
13
2103
NM_018212.4
NP_060682.2
12
2040
XM_005273187.2
XP_005273244.1
14
2143
XM_011544229.2
XP_011542531.2
14
2619
XM_017001746.1
XP_016857235.1
15
3378
XM_017001747.1
XP_016857236.1
14
2568
XM_017001748.1
XP_016857237.1
13
2562
XM_017001749.1
XP_016857238.1
13
2556
XM_017001750.1
XP_016857239.1
14
2520
XM_017001751.1
XP_016857240.1
12
2388
XM_017001752.1
XP_016857241.1
11
2388
XM_017001753.1
XP_016857242.1
16
2182
XM_017001754.1
XP_016857243.1
15
2126
XM_017001755.1
XP_016857244.1
15
1869
XM_017001756.1
XP_016857245.1
13
1800
XM_017001757.1
XP_016857246.1
13
1703
GenBank mRNA
AF519769.1
-
13
1919
AK001635.1
-
6
520
AK096246.1
-
10
1336
AK126894.1
-
7
797
AK127561.1
-
7
1234
AK290179.1
-
12
1984
AL133059.1
-
6
411
AY345143.1
-
12
1944
BC065238.1
-
11
1458
BC095481.1
-
13
2102
30348815
EU255274.1
-
10
1480
KM214203.1
-
12
1644
KM214204.1
-
13
1707
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
axon guidance
actin binding
protein binding
SH3 domain binding
WW domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-x88h5:80/100.66.78.96:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline