Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008493.1 | NP_001008493.1 | 5 | 967 | ||
NM_018212.4 | NP_060682.2 | 5 | 967 | |||
XM_005273187.2 | XP_005273244.1 | 6 | 1007 | |||
XM_011544229.2 | XP_011542531.2 | 6 | 1483 | |||
XM_017001746.1 | XP_016857235.1 | 7 | 2242 | |||
XM_017001747.1 | XP_016857236.1 | 7 | 1495 | |||
XM_017001748.1 | XP_016857237.1 | 5 | 1426 | |||
XM_017001749.1 | XP_016857238.1 | 6 | 1483 | |||
XM_017001750.1 | XP_016857239.1 | 7 | 1495 | |||
XM_017001751.1 | XP_016857240.1 | 5 | 1315 | |||
XM_017001752.1 | XP_016857241.1 | 5 | 1426 | |||
XM_017001753.1 | XP_016857242.1 | 8 | 1046 | |||
XM_017001754.1 | XP_016857243.1 | 7 | 990 | |||
XM_017001755.1 | XP_016857244.1 | 7 | 733 | |||
XM_017001756.1 | XP_016857245.1 | 5 | 664 | |||
XM_017001757.1 | XP_016857246.1 | 5 | 567 | |||
GenBank mRNA | AF519769.1 | - | 5 | 783 | ||
AK096246.1 | - | 3 | 263 | |||
AK290179.1 | - | 5 | 911 | |||
AY345143.1 | - | 5 | 871 | |||
BC065238.1 | - | 4 | 385 | |||
BC095481.1 | - | 5 | 966 | 30348815 | ||
EU255274.1 | - | 4 | 518 | |||
KM214203.1 | - | 5 | 571 | |||
KM214204.1 | - | 5 | 571 |